MilliporeSigma
Magnetic phases of cobalt atomic clusters on tungsten.
Journal of physics. Condensed matter : an Institute of Physics journal (2012-12-12)
Pavel V Lukashev, Ji-Hyun Kim, Seolun Yang, Jae-Sung Kim, Xumin Chen, Geoffrey Rojas, Jan Honolka, Ralph Skomski, Axel Enders, Renat F Sabirianov
ABSTRACT
First-principle calculations are employed to show that the magnetic structure of small atomic clusters of Co, formed on a crystalline W(110) surface and containing 3-12 atoms, strongly deviates from the usual stable ferromagnetism of Co in other systems. The clusters are ferri-, ferro- or non-magnetic, depending on cluster size and geometry. We determine the atomic Co moments and their relative alignment, and show that antiferromagnetic spin alignment in the Co clusters is caused by hybridization with the tungsten substrate and band filling. This is in contrast with the typical strong ferromagnetism of bulk Co alloys, and ferromagnetic coupling in Fe/W(110) clusters.
Sigma-Aldrich
Tungsten, powder, 12 μm, 99.9% trace metals basis
Sigma-Aldrich
Tungsten, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tungsten, powder (monocrystalline), 0.6-1 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Tungsten, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tungsten, powder, ≤10 μm, ≥99.99% trace metals basis
Sigma-Aldrich
Tungsten, foil, thickness 0.127 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tungsten, foil, thickness 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tungsten, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Tungsten, wire, diam. 0.25 mm, ≥99.9% trace metals basis