MilliporeSigma
  • A Raman study on preferential interactions in the formamide/pyridine/pyridazine system and complementary thermodynamic information on the formamide/pyridazine mixture.
A Raman study on preferential interactions in the formamide/pyridine/pyridazine system and complementary thermodynamic information on the formamide/pyridazine mixture.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2012-12-12)
Elaine F da Silva, Wagner A Alves
ABSTRACT
The formamide (FA)/pyridine (py)/pyridazine (prd) system was investigated by FT-Raman spectroscopy and the results in situ show that FA is preferentially bound to py, as experimentally pointed out by the proton affinity (PA) values of these azabenzenes. Temperature-dependent further investigations for the FA/prd mixture clearly show that the 2:1 FA:prd complex is more stable than the 1:1 FA:py adduct, even though its formation is influenced by the extremely negative entropy of the system, which becomes the spontaneously unfavorable global process. Linear relationships of the complex formation enthalpies (ΔH°) with band shifts (Δν) and dipole moments (μ) of these azabenzenes are observed and are in excellent agreement with our recent studies.
Sigma-Aldrich
Formamide, ACS reagent, ≥99.5%
Sigma-Aldrich
Formamide, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Formamide, BioReagent, ≥99.5% (GC), for molecular biology
Sigma-Aldrich
Formamide, puriss. p.a., ACS reagent, ≥99.5% (GC/T)
Sigma-Aldrich
Formamide, spectrophotometric grade, ≥99%
Sigma-Aldrich
Formamide, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Formamide, puriss. p.a., ACS reagent, ≥99.5% (GC/T)
Sigma-Aldrich
Pyridazine, 98%