MilliporeSigma
  • Structural characteristics and antioxidant activities of polysaccharides from longan seed.
Structural characteristics and antioxidant activities of polysaccharides from longan seed.
Carbohydrate polymers (2012-12-12)
Guoxiang Jiang, Lingrong Wen, Feng Chen, Fuwang Wu, Sen Lin, Bao Yang, Yueming Jiang
ABSTRACT
Ultrasound-assisted extraction was employed to extract polysaccharides from longan seed (LSP), with aids of a Box-Behnken statistical design to investigate the effects of ultrasonic power, time and liquid/solid ratio on the extraction recovery of the LSP. The structural analysis indicated that arabinose, galactose, glucose and mannose were major components of LSP, with →6)-Gal-(→1, Glc-(→1 and →6)-Glc-(1→ glycosidic linkages. In an in vitro antioxidant activity of the 1,1-diphenyl-2-picryldydrazyl radical-scavenging assay, LSP exhibited a dose-dependent property within the concentration range tested.
Product Description
Supelco
Biphenyl, PESTANAL®, analytical standard
Sigma-Aldrich
Biphenyl, ≥99%
Sigma-Aldrich
Biphenyl, ReagentPlus®, 99.5%
Sigma-Aldrich
DL-Arabinose, ≥98% (GC)