MilliporeSigma
[Chemical constituents of Pileostegia viburnoides var. glabrescens].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2012-12-12)
Ju-ying Zou, Sheng-huang Chen, Qin-wen Li, Yang-wen Ou, Han-jun Chen, Wei Wang
ABSTRACT
To study the chemical constituents of Pileostegia viburnoides var. glabrescens. The compounds were isolated and purified by various techniques. Their structures were determined by physicochemical properties and spectral analysis. Five compounds were isolated and identified as friedelin (1), beta-sitosterol (2), umbelliferone (3), daucosterol (4) and skimmin (5). All the compounds were isolated from this genus for the first time.
Sigma-Aldrich
Friedelin, technical grade
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Please contact Customer Service for Availability
$619.00
Supelco
β-Sitosterol β-D-glucoside, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mg
Please contact Customer Service for Availability
$819.00
Sigma-Aldrich
β-Sitosterol, from soybean, ≥96%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mg
Please contact Customer Service for Availability
$157.00
10 mg
Please contact Customer Service for Availability
$723.00
Sigma-Aldrich
β-Sitosterol, synthetic, ≥95%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mg
Please contact Customer Service for Availability
$323.00
25 mg
Please contact Customer Service for Availability
$549.00
100 mg
Please contact Customer Service for Availability
$1,590.00
Sigma-Aldrich
Umbelliferone, 99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Umbelliferone, suitable for fluorescence indicator, ≥98.0% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
β-Sitosterol, ≥70%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 g
Please contact Customer Service for Availability
$132.00
Supelco
β-Sitosterol, certified reference material, 100 μg/mL in chloroform
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mL
Please contact Customer Service for Availability
$77.70
Supelco
Umbelliferone, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 mg
Please contact Customer Service for Availability
$181.00