MilliporeSigma
  • Gastro-intestinal transport of calcium and cadmium in fresh water and seawater acclimated trout (Oncorhynchus mykiss).
Gastro-intestinal transport of calcium and cadmium in fresh water and seawater acclimated trout (Oncorhynchus mykiss).
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP (2012-12-04)
Joel S Klinck, Chris M Wood
ABSTRACT
Transport of calcium (Ca) and cadmium (Cd) was examined along the gastro-intestinal tract (GIT) of freshwater and seawater Oncorhynchus mykiss irideus (FWT and SWTies respectively) using in vitro and in vivo experiments. Based on known physiological differences between FWT and SWT which aid in regulating ion levels and osmolarity, we hypothesized that SWT would have lower rates of Ca uptake. Also, we predicted that Cd rates would also be lower because Cd is known to share a common transport mechanism with Ca. Kinetics of Ca and Cd transport were determined using mucosal salines of varying concentrations [1, 10, 30, 60, and 100 (mmolL(-1) for Ca, μmolL(-1) for Cd)]. Linear and saturating relationships were found for Ca for FWT and SWT, but overall SWT had lower rates. Linear and/or saturating relationships were also found for Cd uptake, but rates varied little between fish types. Elevated Ca had no inhibitory effect on Cd transport, and Ca channel blockers nifedipine and verapamil had little effect on Ca or Cd uptake. However, lanthanum reduced Ca transport into some compartments. A 21 day in vivo feeding experiment was also performed where FWT and SWT were exposed to control diets or Cd-spiked diets (552 μg Cd g(-1) food). Whole body Cd uptake between fish types was similar, but the majority of Cd in SWT remained in the posterior intestine tissue, while FWT transported more Cd through their gut wall. Overall it appears that large differences in Ca and Cd uptake between FWT and SWT exist, with SWT generally having lower rates.
Sigma-Aldrich
Nifedipine, ≥98% (HPLC), powder
Nifedipine, European Pharmacopoeia (EP) Reference Standard