MilliporeSigma
  • Increased apoptosis in osteoclasts and decreased RANKL immunoexpression in periodontium of cimetidine-treated rats.
Increased apoptosis in osteoclasts and decreased RANKL immunoexpression in periodontium of cimetidine-treated rats.
Journal of anatomy (2012-12-04)
Renata Longhini, Priscila Aparecida de Oliveira, Ana Paula de Souza Faloni, Estela Sasso-Cerri, Paulo Sérgio Cerri
ABSTRACT
It has been demonstrated that histamine interferes with the recruitment, formation and activity of osteoclasts via H(1)- and H(2)-receptors. Cimetidine is a H(2)-receptor antagonist used for treatment of gastric ulcers that seems to prevent bone resorption. In this study, a possible cimetidine interference was investigated in the number of alveolar bone osteoclasts. The incidence of osteoclast apoptosis and immunoexpression of RANKL (receptor activator of nuclear factor κB ligand) was also evaluated. Adult male rats were treated with 100 mg kg(-1) of cimetidine for 50 days (CimG); the sham group (SG) received saline. Maxillary fragments containing the first molars and alveolar bone were fixed, decalcified and embedded in paraffin. The sections were stained by H&E or submitted to tartrate-resistant acid phosphatase (TRAP) method. TUNEL (terminal deoxynucleotidyl transferase-mediated dUTP nick-end labeling) method and immunohistochemical reactions for detecting caspase-3 and RANKL were performed. The number of TRAP-positive osteoclasts, the frequency of apoptotic osteoclasts and the numerical density of RANKL-positive cells were obtained. Osteoclast death by apoptosis was confirmed by transmission electron microscopy (TEM). In CimG, TRAP-positive osteoclasts with TUNEL-positive nuclei and caspase-3-immunolabeled osteoclasts were found. A significant reduction in the number of TRAP-positive osteoclasts and a high frequency of apoptotic osteoclasts were observed in CimG. Under TEM, detached osteoclasts from the bone surface showed typical features of apoptosis. Moreover, a significant reduction in the numerical density of RANKL-positive cells was observed in CimG. The significant reduction in the number of osteoclasts may be due to cimetidine-induced osteoclast apoptosis. However, RANKL immunoexpression reduction also suggests a possible interference of cimetidine treatment in the osteoclastogenesis.
Product Description
Supelco
Cimetidine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Cimetidine