MilliporeSigma
  • Chemoselective synthesis of substituted pyrazoles through AgOTf-catalyzed cascade propargylic substitution-cyclization-aromatization.
Chemoselective synthesis of substituted pyrazoles through AgOTf-catalyzed cascade propargylic substitution-cyclization-aromatization.
Organic & biomolecular chemistry (2012-11-29)
Su-Xia Xu, Lu Hao, Tao Wang, Zong-Cang Ding, Zhuang-Ping Zhan
ABSTRACT
A cascade AgOTf-catalyzed chemoselective approach to 3,5/1,3-disubstitued pyrazoles from propargylic alcohols and para-tolylsulfonohydrazide has been developed. Good chemoselectivity is observed depending on the different substituents in the alkyne moiety of the propargylic alcohols, generating two different kinds of products through different aromatization mechanisms. The pyrazolo[5,1-a]isoquinoline skeleton can also be effectively constructed by this method through a cascade bicyclization process.
Sigma-Aldrich
Lanthanum(III) trifluoromethanesulfonate, 99.999% trace metals basis
Sigma-Aldrich
Trifluoromethanesulfonic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Lithium trifluoromethanesulfonate, 96%
Sigma-Aldrich
Trifluoromethanesulfonic acid, reagent grade, 98%
Sigma-Aldrich
Propargyl alcohol, 99%
Sigma-Aldrich
Lithium trifluoromethanesulfonate, 99.995% trace metals basis
Sigma-Aldrich
Zinc trifluoromethanesulfonate, 98%
Sigma-Aldrich
Silver trifluoromethanesulfonate, purum, ≥98.0% (Ag)
Sigma-Aldrich
Silver trifluoromethanesulfonate, ≥99.95% trace metals basis
Sigma-Aldrich
Aluminum trifluoromethanesulfonate, 99.9% trace metals basis
Sigma-Aldrich
Barium trifluoromethanesulfonate, 98%
Sigma-Aldrich
Silver trifluoromethanesulfonate, ≥99%