MilliporeSigma
Laser action in Nd(3+)-doped lanthanum oxysulfide powders.
Optics express (2012-11-29)
Iñaki Iparraguirre, Jon Azkargorta, Odile Merdrignac-Conanec, Mohamad Al-Saleh, Christophe Chlique, Xianghua Zhang, Rolindes Balda, Joaquín Fernández
ABSTRACT
We have investigated the stimulated emission properties of Nd(3+) doped La(2)O(2)S powders at room temperature as a function of pumping energy density, excitation wavelength, and Nd(3+) ion concentration. The absolute stimulated emission energy has been measured. Expressions for the slope efficiencies and lasing thresholds as a function of rare earth concentration and pumping wavelengths, which qualitatively agree with experimental observations, are discussed.
Sigma-Aldrich
Lanthanum(III) oxide, 99.99% trace metals basis
Sigma-Aldrich
Neodymium, powder, −40 mesh, ≥99% trace rare earth metals basis
Supelco
Lanthanum(III) oxide, suitable for AAS, ≥99.9%
Sigma-Aldrich
Lanthanum(III) oxide, 99.999% trace metals basis
Sigma-Aldrich
Lanthanum(III) oxide, ≥99.9%
Sigma-Aldrich
Lanthanum(III) oxide, nanopowder, <100 nm particle size (TEM), 99% trace metals basis