MilliporeSigma
Adherence of Streptococcus mutans to orthodontic band cements.
Australian dental journal (2012-11-29)
J C Gonzalez-Perez, R J Scougall-Vilchis, R Contreras-Bulnes, I De La Rosa-Gómez, S Uematsu, R Yamaguchi
ABSTRACT
The aim of this study was to quantitatively determine the adherence of Streptococcus mutans to orthodontic band cements. Two hundred and ten blocks of seven different band cements for orthodontic prescription were made using a Teflon mould (4 × 4 × 1 mm). The obtained blocks were slightly polished and cleansed ultrasonically. Certified S. mutans ATCC 25175 were cultured with conventional methods for growth in Petri dishes and trypticase soy broth. Quantitative analysis was carried out with radioactive markers to codify the bacteria ((3) H). Subsequently, a combustion system was used to capture the residues, the radioactivity of the samples was measured, and the values recorded in disintegrations per minute (dpm). One-way analysis of variance (ANOVA) with a Scheffé test for multiple comparisons was realized with a significance level of ≤0.05. Significant differences were found among different band cement materials (p < 0.001). Two band cement materials showed statistically lower values than the others (Transbond Plus Band Cem and Ketac Cem). In contrast, GC Fuji Ortho Band presented the highest adherence of S. mutans. Among the cements evaluated, Transbond Plus Band Cem and Ketac Cem showed lower adherence of S. mutans.
Sigma-Aldrich
Magnesium oxide, tested according to Ph. Eur., heavy
Sigma-Aldrich
Magnesium oxide, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium oxide, nanopowder, ≤50 nm particle size (BET)
Sigma-Aldrich
Magnesium oxide, BioUltra, ≥97.0% (calcined substance, KT)
Sigma-Aldrich
Magnesium oxide, 99.99% trace metals basis
Sigma-Aldrich
Magnesium oxide, (single crystal substrate), ≥99.9% trace metals basis, <100>, L × W × thickness 10 mm × 10 mm × 0.5 mm
Sigma-Aldrich
Magnesium oxide, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 530, light, 98.0-100.5% (calc. for dried substance)
Sigma-Aldrich
Magnesium oxide, -10-+50 mesh, 98%
Sigma-Aldrich
Magnesium oxide, ACS reagent, 97%
Sigma-Aldrich
Magnesium oxide, light, 95%
Sigma-Aldrich
Magnesium oxide, ≥99% trace metals basis, -325 mesh