MilliporeSigma
  • [Biological markers for the status of vitamins B12 and D: the importance of some analytical aspects in relation to clinical interpretation of results].
[Biological markers for the status of vitamins B12 and D: the importance of some analytical aspects in relation to clinical interpretation of results].
Revue medicale suisse (2012-11-29)
O Boulat, F Rey, V Mooser
ABSTRACT
Biological markers for the status of vitamins B12 and D: the importance of some analytical aspects in relation to clinical interpretation of results When vitamin B12 deficiency is expressed clinically, the diagnostic performance of total cobalamin is identical to that of holotranscobalamin II. In subclinical B12 deficiency, the two aforementioned markers perform less well. Additional analysis of a second, functional marker (methylmalonate or homocysteine) is recommended. Different analytical approaches for 25-hydroxyvitamin D quantification, the marker of vitamin D deficiency, are not yet standardized. Measurement biases of up to +/- 20% compared with the original method used to establish threshold values are still observed.
Sigma-Aldrich
Methylmalonic acid, 99%
Supelco
Methylmalonic acid solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®