MilliporeSigma
  • Size-controlled and biocompatible Gd2 O3 nanoparticles for dual photoacoustic and MR imaging.
Size-controlled and biocompatible Gd2 O3 nanoparticles for dual photoacoustic and MR imaging.
Advanced healthcare materials (2012-11-28)
Yu Kimura, Ryota Kamisugi, Michiko Narazaki, Tetsuya Matsuda, Yasuhiko Tabata, Akio Toshimitsu, Teruyuki Kondo
ABSTRACT
The synthesis, characterization, and functional evaluation of new size-controlled and biocompatible Gd(2) O(3) nanoparticles as a bimodal contrast agent for use in photoacoustic (PA) and magnetic resonance (MR) imaging are reported. These nanoparticles show a clear PA image by themselves, without conjugation with gold, rare earth metals, or dyes. Relaxivity measurement by MR imaging clearly shows that their relaxivity, r(1) , is twice that of clinically available Gd-DTPA.
Sigma-Aldrich
Gadolinium(III) oxide, ≥99.9%
Sigma-Aldrich
Gadolinium(III) oxide, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Gadolinium(III) oxide, powder, 99.9% trace metals basis
Sigma-Aldrich
Gadolinium(III) oxide, nanopowder, <100 nm particle size (BET), 99.8% trace metals basis