MilliporeSigma
  • Cell-specific intracellular anticancer drug delivery from mesoporous silica nanoparticles with pH sensitivity.
Cell-specific intracellular anticancer drug delivery from mesoporous silica nanoparticles with pH sensitivity.
Advanced healthcare materials (2012-11-28)
Zhong Luo, Kaiyong Cai, Yan Hu, Beilu Zhang, Dawei Xu
ABSTRACT
A nanoreservoir for efficient intracellular anticancer drug delivery based on mesoporous silica nanoparticles end-capped with lactobionic acid-grafted bovine serum albumin is fabricated. It demonstrates great potential for both cell-specific endocytosis and intracellular pH-responsive controlled release of drugs. A possible endocytosis pathway/mechanism of the smart controlled drug release system is proposed.
Sigma-Aldrich
Lactobionic acid, ≥97% (TLC), cell impermeant agent
Sigma-Aldrich
Lactobionic acid, 97% (TLC)
Lactobionic acid, European Pharmacopoeia (EP) Reference Standard