MilliporeSigma
Venlafaxine and neuropathic pain.
Pharmacology (2012-11-28)
Krystyna Cegielska-Perun, Magdalena Bujalska-Zadrożny, Jan Tatarkiewicz, Emilia Gąsińska, Helena Elżbieta Makulska-Nowak
ABSTRACT
The possible mechanisms involved in the antinociceptive effect of venlafaxine (VFX), a selective serotonin and noradrenaline reuptake inhibitor, after a single administration and chronic treatment were investigated in a diabetic neuropathic pain (DNP) model. VFX produced a significant antihyperalgesic effect after a single and repeated administration. This effect was reversed by pretreatment with yohimbine (a relatively selective α(2)-adrenergic antagonist) and p-chloroamphetamine (a neurotoxin which destroys serotonergic neurons). Conversely, naloxone (a nonselective opioid antagonist) did not reverse the effect of VFX in a DNP model. It is concluded that both noradrenergic and serotonergic mechanisms participate in the antinociceptive effect of VFX in the DNP model. However, the noradrenergic mechanism probably plays a more important role.
Sigma-Aldrich
Venlafaxine hydrochloride, ≥98% (HPLC), powder
Supelco
Venlafaxine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Venlafaxine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Venlafaxine for system suitability, European Pharmacopoeia (EP) Reference Standard