MilliporeSigma
  • Suppression of MAPK and NF-κB pathways by limonene contributes to attenuation of lipopolysaccharide-induced inflammatory responses in acute lung injury.
Suppression of MAPK and NF-κB pathways by limonene contributes to attenuation of lipopolysaccharide-induced inflammatory responses in acute lung injury.
Inflammation (2012-11-28)
Gefu Chi, Miaomiao Wei, Xianxing Xie, L W Soromou, Fang Liu, Shuhua Zhao
ABSTRACT
The present study aimed to investigate the protective role of limonene in lipopolysaccharide (LPS)-induced acute lung injury (ALI). ALI was induced in mice by intratracheal instillation of LPS (0.5 mg/kg), and limonene (25, 50, and 75 mg/kg) was injected intraperitoneally 1 h prior to LPS administration. After 12 h, bronchoalveolar lavage fluid (BALF) and lung tissue were collected. Limonene pretreatment at doses of 25, 50, and 75 mg/kg decreased LPS-induced evident lung histopathological changes, lung wet-to-dry weight ratio, and lung myeloperoxidase activity. In addition, pretreatment with limonene inhibited inflammatory cells and proinflammatory cytokines including tumor necrosis factor-α, interleukin-1β, and interleukin-6 in BALF. Furthermore, we demonstrated that limonene blocked the phosphorylation of IκBα, nuclear factor-κB (NF-κB) p65, p38 mitogen-activated protein kinase (MAPK), c-Jun NH2-terminal kinase, and extracellular signal-regulated kinase in LPS-induced ALI. The results presented here suggest that the protective mechanism of limonene may be attributed partly to decreased production of proinflammatory cytokines through the inhibition of NF-κB and MAPK activation.
Sigma-Aldrich
(R)-(+)-Limonene, 97%
Sigma-Aldrich
(R)-(+)-Limonene, technical, ~90% (sum of enantiomers, GC)
Supelco
(R)-(+)-Limonene, analytical standard
Sigma-Aldrich
(S)-(−)-Limonene, 96%
Sigma-Aldrich
(S)-(−)-Limonene, ≥95%, FG
Supelco
(S)-(−)-Limonene, analytical standard