MilliporeSigma
  • Orofacial dyskinesia induced by nasal Ritalin(R) (methylphenidate) sniffing: a rare case report from Switzerland.
Orofacial dyskinesia induced by nasal Ritalin(R) (methylphenidate) sniffing: a rare case report from Switzerland.
Human & experimental toxicology (2012-11-24)
G Marti, K Fattinger, H Zimmermann, A Exadaktylos
ABSTRACT
Ritalin® (methylphenidate) is an amphetamine-like prescription stimulant commonly used in the treatment of attention deficit hyperactivity disorder in children and adults. Recently, the recreational use of Ritalin has increased, particularly among young adults. Well-known symptoms of intoxication include signs of sympathetic nervous stimulation, such as agitation, anxiety, tachycardia, hypertension, headache, tremor, and dizziness. This case report describes oral dyskinesia as a rare presentation of Ritalin intoxication, with the review of pathophysiology and some epidemiological data.
Product Description
Supelco
Lorazepam solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®