MilliporeSigma
  • Amyloid β-peptide 25-35 self-assembly and its inhibition: a model undecapeptide system to gain atomistic and secondary structure details of the Alzheimer's disease process and treatment.
Amyloid β-peptide 25-35 self-assembly and its inhibition: a model undecapeptide system to gain atomistic and secondary structure details of the Alzheimer's disease process and treatment.
ACS chemical neuroscience (2012-11-23)
Marina Naldi, Jessica Fiori, Marco Pistolozzi, Alex F Drake, Carlo Bertucci, Rongliang Wu, Krzysztof Mlynarczyk, Slawomir Filipek, Angela De Simone, Vincenza Andrisano
ABSTRACT
Combined results of theoretical molecular dynamic simulations and in vitro spectroscopic (circular dichroism and fluorescence) studies are presented, providing the atomistic and secondary structure details of the process by which a selected small molecule may destabilize the β-sheet ordered "amyloid" oligomers formed by the model undecapeptide of amyloid β-peptide 25-35 [Aβ(25-35)]. Aβ(25-35) was chosen because it is the shortest fragment capable of forming large β-sheet fibrils and retaining the toxicity of the full length Aβ(1-40/42) peptides. The conformational transition, that leads to the formation of β-sheet fibrils from soluble unordered structures, was found to depend on the environmental conditions, whereas the presence of myricetin destabilizes the self-assembly and antagonizes this conformational shift. In parallel, we analyzed several molecular dynamics trajectories describing the evolution of five monomer fragments, without inhibitor as well as in the presence of myricetin. Other well-known inhibitors (curcumin and (-)-tetracycline), found to be stronger and weaker Aβ(1-42) aggregation inhibitors, respectively, were also studied. The combined in vitro and theoretical studies of the Aβ(25-35) self-assembly and its inhibition contribute to understanding the mechanism of action of well-known inhibitors and the peptide amino acid residues involved in the interaction leading to a rational drug design of more potent new molecules able to antagonize the self-assembly process.
Sigma-Aldrich
Myricetin, ≥96.0% (HPLC)
Sigma-Aldrich
Amyloid β-Protein Fragment 25-35, ≥97% (HPLC)
Sigma-Aldrich
Myricetin, ≥96.0%, crystalline
Supelco
Myricetin, analytical standard