MilliporeSigma
  • Type-II nanorod heterostructure formation through one-step cation exchange.
Type-II nanorod heterostructure formation through one-step cation exchange.
Nanoscale (2012-11-23)
Meng-Yu Chen, Yung-Jung Hsu
ABSTRACT
A novel one-step cation exchange approach has been developed to prepare ZnO-decorated ZnSe nanorods (ZnSe-ZnO NRs), a prototype type-II semiconductor nanoheterostructure. Because of the staggered band offset which promoted effective charge separation, the as-synthesized ZnSe-ZnO NRs exhibited remarkable photocatalytic activities under visible light illumination, demonstrating their promising potentials in relevant photoconversion applications.
Sigma-Aldrich
Zinc selenide, powder, 10 μm, 99.99% trace metals basis