MilliporeSigma
  • Influence of gum tragacanth on the physicochemical and rheological properties of kashk.
Influence of gum tragacanth on the physicochemical and rheological properties of kashk.
The Journal of dairy research (2012-11-23)
Setareh Ghorban Shiroodi, Mohammad Amin Mohammadifar, Elham Ghorbani Gorji, Hamid Ezzatpanah, Nilofar Zohouri
ABSTRACT
In this study, the physicochemical properties of a low-fat dried yogurt paste (kashk) were determined, and the effects of different concentrations (0, 0·1, 0·3 and 0·5% w/w) of gum tragacanth exudates from Astragalus gossypinus on the stability and texture of the samples were investigated by measuring amount of syneresis, turbidity, particle size distribution (PSD), flow behaviour and viscoelastic properties. The flow behaviour index was not very sensitive to the concentration of gum, while a remarkable concentration dependency of the power-law consistency coefficient and Herschel-Bulkley yield stress was observed. The initial increase in the gum concentration at 0·1 and 0·3% levels led to a higher degree of syneresis, which was related to the depletion flocculation mechanism. However, the reduced amount of syneresis in samples containing 0·5% gum tragacanth was attributed to the significant increase in viscosity of the continuous phase, which is also accompanied by trapping of the aggregated casein particles. The presence of 3% salt in the samples may have led to the neutralization of charges on the surface of gum tragacanth; consequently, the non-adsorbing behaviour of high-ionic-strength polysaccharides inhibited the formation of electrostatic protein-polysaccharide complexes. Furthermore, maximum values of polydispersity, syneresis and tan δ at high frequencies were found in samples containing 0·1% gum tragacanth.
Sigma-Aldrich
Tragacanth