MilliporeSigma
  • Nanostructural transformations during the reduction of hollow and porous nickel oxide nanoparticles.
Nanostructural transformations during the reduction of hollow and porous nickel oxide nanoparticles.
Nanoscale (2012-11-22)
John A Medford, Aaron C Johnston-Peck, Joseph B Tracy
ABSTRACT
Size-dependent nanostructural transformations occurring during the H(2)-mediated reduction of hollow and porous NiO nanoparticles were investigated for controlled nanoparticle sizes of ~10 to 100 nm. Transmission electron microscopy reveals that the location and number of reduction sites strongly depend on the nanoparticle size and structure.
Sigma-Aldrich
Nickel(II) oxide, nanopowder, <50 nm particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Nickel(II) oxide, 99.99% trace metals basis
Sigma-Aldrich
Nickel(II) oxide, green, −325 mesh, 99%
Sigma-Aldrich
Nickel(II) oxide, ≥99.995% trace metals basis