MilliporeSigma
  • ZnO/TiO2 nanocable structured photoelectrodes for CdS/CdSe quantum dot co-sensitized solar cells.
ZnO/TiO2 nanocable structured photoelectrodes for CdS/CdSe quantum dot co-sensitized solar cells.
Nanoscale (2012-11-21)
Jianjun Tian, Qifeng Zhang, Lili Zhang, Rui Gao, Laifa Shen, Shengen Zhang, Xuanhui Qu, Guozhong Cao
ABSTRACT
Photoelectrode made of nanocable structure of ZnO nanorods (NR) coated with TiO(2) nanosheets (NSs) was investigated for CdS/CdSe quantum dot co-sensitized solar cells. ZnO NRs prepared solution reaction at 60 °C served as the backbone for direct electron transport in view of the single crystallinity of the ZnO NRs and the high electron mobility of ZnO semiconductor. Anatase TiO(2) NSs with the thickness of ∼10 nm and the length of ∼100 nm were assembled onto the surface of ZnO NRs via a solvothermal method. It was found that the thin shell of TiO(2) might have remarkable influence on the quantum dot sensitized solar cells (QDSCs) through (a) increasing the surface area of ZnO NRs to allow for adsorbing more quantum dots (QDs), which led to high short current density, (b) forming an energy barrier that hindered the electrons in the ZnO from being back to the electrolyte and QDs, and thus, reduced the charge recombination rate, resulting in prolonged electron lifetime and enhanced open voltage. In comparison with the case of ZnO NRs, the short-circuit current density, open-circuit voltage, fill factor and charge recombination resistance of ZnO/TiO(2) nanocable photoelectrode increase by 3%, 44%, 48% and 220%, respectively. As a result, a power conversion efficiency of 2.7% of QDSCs with core-shell structural nanocable photoelectrode has been obtained, which is as much as 230% of that of 1.2% obtained for ZnO NR photoelectrode.