MilliporeSigma
Photoluminescence of silicon quantum dots in nanospheres.
Nanoscale (2012-11-10)
Yuxiao Zhang, Xiao Han, Jianming Zhang, Yang Liu, Hui Huang, Hai Ming, Shuit-Tong Lee, Zhenhui Kang
ABSTRACT
Si quantum dots (SiQDs) based nanospheres (SiNSs) were prepared via a novel synthetic strategy. These SiNSs were demonstrated to possess unique dot spacing dependent photoluminescence (PL) up-conversion and surface dependent (N modified surface) down-converted PL. It was demonstrated that a small distance between SiQDs (<5 nm) is the necessary condition for the PL up-conversion of SiNSs, while the surface state of SiQDs will affect the maximum emission wavelength and the PL intensity. The as-prepared SiNSs feature excellent aqueous dispersibility, and their optical properties were found to be stable enough in a specified temperature and pH range.
Sigma-Aldrich
Acrylic acid, anhydrous, contains 200 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Zinc acrylate, 98%
Sigma-Aldrich
Ethylenediamine dihydrochloride, 98%
Sigma-Aldrich
Ethylenediamine solution, technical, 75-80%
Supelco
Ethylenediamine, analytical standard
Sigma-Aldrich
Ethylenediamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
Ethylenediamine, puriss. p.a., absolute, ≥99.5% (GC)
Sigma-Aldrich
Ethylenediamine, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethylenediamine, BioXtra
Sigma-Aldrich
Ethylenediamine, meets USP testing specifications