MilliporeSigma
  • Cytogenetic evaluation and the association with polymorphisms of the CPY1A1 and NR1I3 genes in individuals exposed to BTEX.
Cytogenetic evaluation and the association with polymorphisms of the CPY1A1 and NR1I3 genes in individuals exposed to BTEX.
Environmental monitoring and assessment (2012-11-10)
João Carlos Fraga da Rosa, Marilu Fiegenbaum, Ane Lise Soledar, Matheus Souza Claus, Antonio Daniel de Souza Nunes, Valesca Veiga Cardoso
ABSTRACT
The gas station attendants are exposed daily to chemical agents that compose gasoline, such as BTEX (benzene, toluene, ethylbenzene, and xylene), and the exposure to these agents can cause a variety of effects on the human health. Among the various possible cell alterations associated with these exposures are the formation of micronuclei and of binucleated cells which are used as indicators of clastogenic action. Benzene, the main carcinogenic agent, is metabolized to more soluble forms and easily excreted by isoenzymes of cytochrome P450, such as CYP1A1. The CYP1A1 gene is highly polymorphic and one of its allele variations can be detected by the use of restriction endonucleasis MspI and is originated by the transition of a thymine by a cytosine (3798T>C), resulting in the polymorphic allele CYP1A1*2A. The objective of this study was to evaluate the cytogenetic damage induced by the exposure to BTEX and to associate it with the polymorphisms of the CYP1A1 and NR1I3 genes. Samples of exfoliated cells from the oral mucosa of 27 gas station attendants and from a control group were collected. The results found show that the group exposed to BTEX presents significantly higher alterations than those in the control group for micronuclei (MN; 6.85 ± 1.33 vs. 2.96 ± 1.91, P < 0.001) and for the total of nuclear alterations observed (MN + binucleated cells (BNC); 9.59 ± 4.73 vs. 5.07 ± 2.21, P < 0.001). When comparing the cytological alterations and the genotypes among the exposed individuals for the polymorphism 3798T>C of the CYP1A1 gene, homozygotes TT present MN + BNC significantly higher than carriers of the allele C (10.88 ± 5.36 vs. 5.33 ± 2.52, P = 0.028). No association was observed in the control group or for the NR1I3 gene. These results show that molecular and cytogenetic data can be used in the future as tools to monitor individuals exposed to such compounds.
Product Description
Supelco
Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), chromium(III) acetylacetonate 0.1 mg/mL, TMS 0.01 %, NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 10 mm × 8 in.
Supelco
Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Ethylbenzene solution, NMR reference standard, 5% in chloroform-d (99.8 atom % D), TMS 2 %, NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Ethylbenzene solution, NMR reference standard, 1% in chloroform-d (99.8 atom % D)
Sigma-Aldrich
Ethylbenzene solution, NMR reference standard, 10% in chloroform-d (99.8 atom % D)
Supelco
Ethylbenzene solution, NMR reference standard, 10% in chloroform-d (99.8 atom % D), NMR tube size 5 mm × 8 in. , ultra-thin wall
Sigma-Aldrich
Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d ("100%", 99.96 atom % D)
Sigma-Aldrich
Ethylbenzene solution, NMR reference standard, 5% in chloroform-d (99.8 atom % D), TMS 1 %, NMR tube size 5 mm × 7 in.
Sigma-Aldrich
Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 5 mm × 8 in. , ultra-thin wall
Sigma-Aldrich
Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), TMS 0.1 %
Sigma-Aldrich
Ethylbenzene solution, NMR reference standard, 10% in chloroform-d (99.8 atom % D), NMR tube size 10 mm × 8 in.
Sigma-Aldrich
Ethylbenzene solution, NMR reference standard, 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 8 mm × 8 in.
Supelco
Ethylbenzene, analytical standard
Sigma-Aldrich
Ethylbenzene, anhydrous, 99.8%
Sigma-Aldrich
Ethylbenzene, ReagentPlus®, 99%