MilliporeSigma
  • The relative effects of fat versus muscle mass on cystatin C and estimates of renal function in healthy young men.
The relative effects of fat versus muscle mass on cystatin C and estimates of renal function in healthy young men.
Annals of clinical biochemistry (2012-11-07)
Janice S C Chew-Harris, Christopher M Florkowski, Peter M George, Jane L Elmslie, Zoltan H Endre
ABSTRACT
It is well known that plasma creatinine concentration is affected by muscle mass, while some studies have suggested cystatin C is affected by body mass index (BMI). Our aim was to assess the effects of lean versus fat mass on cystatin C and creatinine derivative equations in estimating glomerular filtration rate (GFR) in healthy young men. Three groups of participants were studied: those classified as normal (BMI 18-25 kg/m(2) with body fat <30%); muscular subjects (BMI >30 kg/m(2) and body fat <20%); and obese subjects (BMI >30 kg/m(2) and body fat >30%). All underwent diethylenetriamine pentaacetic acid GFR, bio-electrical impedance and dual-energy X-ray absorptiometry body composition analysis, measurement of plasma cystatin C, creatinine and high-sensitivity C-reactive protein and completed a diet record. Cystatin C was highest in the obese group (0.77 mg/L; 95% confidence intervals [CI] 0.69-0.77) and creatinine was highest in the muscular group (90.1 μmol/L; 95% CI 84.3-96.0). On multivariate analysis, body fat and GFR (P = 0.003) were significant determinants of cystatin C; muscle mass and age affected creatinine significantly (P = 0.02). Using cystatin C equations, Le Bricon and Hoek showed significantly lower estimated GFR in the obese group but performed reasonably well within 50%, 30% and 20% of GFR. Creatinine equations showed significant underestimations of GFR for the muscular group. Body fat is a significant determinant of cystatin C while creatinine concentration is highly affected by muscle mass and age. Body composition plays an important role in the interpretation of renal function. Cystatin C equations are still accurate in predicting GFR in our healthy male group without chronic kidney disease.
Sigma-Aldrich
Diethylenetriaminepentaacetic acid, ≥98% (titration)
Sigma-Aldrich
Diethylenetriaminepentaacetic acid, ≥99% (titration)
Sigma-Aldrich
Diethylenetriaminepentaacetic acid, ≥99.0%