MilliporeSigma
  • Evaluation of triclosan and triclocarban at river basin scale using monitoring and modeling tools: implications for controlling of urban domestic sewage discharge.
Evaluation of triclosan and triclocarban at river basin scale using monitoring and modeling tools: implications for controlling of urban domestic sewage discharge.
Water research (2012-11-07)
Jian-Liang Zhao, Qian-Qian Zhang, Feng Chen, Li Wang, Guang-Guo Ying, You-Sheng Liu, Bin Yang, Li-Jun Zhou, Shan Liu, Hao-Chang Su, Rui-Quan Zhang
ABSTRACT
Triclosan (TCS) and triclocarban (TCC) are two commonly used personal care products. They may enter into aquatic environments after consumption and pose potential risks to aquatic organisms. We investigated the occurrence and fate of TCS and TCC in five large rivers (the Liao River, Hai River, Yellow River, Zhujiang River and Dongjiang River) in China, and compared the monitoring data with the predicted results from Level III fugacity modeling. TCS and TCC were detected in the five large rivers with the detection frequencies of 100% or close to 100% in surface water and sediments of almost every river. TCS and TCC were found at concentrations of up to 478 ng/L and 338 ng/L in surface water, and up to 1329 ng/g and 2723 ng/g in sediments. Cluster analysis indicated that the sites with higher concentrations were usually located in or near urban area. Meanwhile, principal component analysis also suggested that the mass inventories of TCS and TCC in water and sediment were significantly influenced by the factors such as the total or untreated urban domestic sewage discharge at river basin scale. The concentrations and mass inventories from the fugacity modeling were found at the same order of magnitude with the measured values, suggesting that the fugacity modeling can provide a useful tool for evaluating the fate of TCS and TCC in riverine environments. Both monitoring and modeling results indicated that the majority of mass inventories of TCS and TCC were stored into sediment, which could be a potential pollution source for river water. The wide presence of TCS and TCC in these large rivers of China implies that better controlling of urban domestic sewage discharge is needed.
Sigma-Aldrich
3,4,4′-Trichlorocarbanilide, 99%