MilliporeSigma
  • Porous organic cage crystals: characterising the porous crystal surface.
Porous organic cage crystals: characterising the porous crystal surface.
Chemical communications (Cambridge, England) (2012-11-06)
Michael J Bojdys, Tom Hasell, Nikolai Severin, Kim E Jelfs, Jürgen P Rabe, Andrew I Cooper
ABSTRACT
The characterisation of porous crystalline solids often relies on single crystal X-ray diffraction, which does not give direct information about the surface of the material. Here, crystals of a porous organic cage, CC3-R, are investigated by atomic force microscopy and shown to possess two distinct gas-solid interfaces, proving that the bulk crystal structure is preserved at the porous crystal surface.
Sigma-Aldrich
1,2-Diaminocyclohexane, mixture of cis and trans, 99%
Sigma-Aldrich
(1S,2S)-(+)-1,2-Diaminocyclohexane, 98%
Sigma-Aldrich
(1R,2R)-(−)-1,2-Diaminocyclohexane, 98%
Sigma-Aldrich
(±)-trans-1,2-Diaminocyclohexane, 99%