MilliporeSigma
  • Sprague-Dawley and Fischer female rats differ in acute effects of fluoxetine on sexual behavior.
Sprague-Dawley and Fischer female rats differ in acute effects of fluoxetine on sexual behavior.
The journal of sexual medicine (2012-11-01)
Chandra Suma J Miryala, Cindy Hiegel, Lynda Uphouse
ABSTRACT
The selective serotonin reuptake inhibitor (SSRI), fluoxetine, leads to sexual dysfunction in a substantial proportion of women. In studies with the Fischer inbred rat, the 5-HT(1A) receptor has been implicated in this sexual dysfunction. Whether this association with 5-HT(1A) receptors holds for other rat strains is not known. The effects of acute fluoxetine on sexual behavior in two strains of rats that differ in their response to a 5-HT(1A) receptor agonist were examined. Whether the strain difference is comparable in naturally cycling and hormonally primed, ovariectomized rats was determined. Proestrous rats and ovariectomized rats, hormonally primed with estradiol benzoate and progesterone, were treated with varying doses of fluoxetine. Sexual behavior was examined before and after treatment with the SSRI. Lordosis to mount ratios, lordosis quality, and proceptive behaviors were quantified. Sprague-Dawley and Fischer females were compared on each of these measures. The IC(50) for inhibition of lordosis behavior was determined. In both the intact and the hormonally primed, ovariectomized model, Sprague-Dawley females were less sensitive to the effects of fluoxetine on sexual behavior. In both groups, fluoxetine showed dose dependency in behavioral inhibition, but a higher dose was required for Sprague-Dawley than for Fischer females. Naturally cycling, proestrous rats required a higher dose of fluoxetine than hormonally primed ovariectomized rats to produce significant inhibition of sexual behavior. Thus, the strain difference in the response to fluoxetine does not parallel strain differences in the response to a 5-HT(1A) receptor agonist. Acute treatment with fluoxetine inhibits lordosis behavior in both Fischer and Sprague-Dawley females and the strain difference cannot be explained by reported strain differences in the response to a 5-HT(1A) receptor agonist. Fluoxetine's inhibition of female rat sexual behavior may involve effects of the SSRI in addition to activation of the 5-HT(1A) receptor.
Sigma-Aldrich
β-Estradiol 3-benzoate, ≥97%