MilliporeSigma
  • [Herpetic infection in patients with psoriasis: the improvement of therapy].
[Herpetic infection in patients with psoriasis: the improvement of therapy].
Klinicheskaia meditsina (2012-10-30)
A A Shul'diakov, T S Barkhatova, S A Satarova, T A Perminova
ABSTRACT
The aim of the study was to estimate the efficacy of liniment cycloferon included in combined therapy of herpetic infection in 30 patients with psoriasis divided into 2 groups. Combined treatment of patients with recurrent herpetic infection promoted elimination of general infection syndrome, shortened duration of eruption and local inflammation, accelerated epithelization of herpetic erosion, and decreased the frequency of relapses during the follow-up.
Sigma-Aldrich
9-Oxo-10(9H)-acridineacetic acid, for HPLC derivatization, ≥99.0% (T)