MilliporeSigma
  • Influence of trace elements on stabilization of aqueous solutions of ascorbic acid.
Influence of trace elements on stabilization of aqueous solutions of ascorbic acid.
Biological trace element research (2012-10-27)
Barbara Dolińska, Aneta Ostróżka-Cieślik, Artur Caban, Klimas Rimantas, Lucyna Leszczyńska, Florian Ryszka
ABSTRACT
Together with vitamin C, zinc, selenium, manganese, and magnesium play a vital role in the preservation of organs scheduled for transplantation. In the present study, it is shown that addition of 1 mg/l of these elements influences the stability of 0.3 mM ascorbic acid solutions. The solution's stability was estimated using an accelerated stability test. The concentration of vitamin C was measured using a validated spectrophotometric method, which uses the reduction of 2,6-dichlorophenoloindophenol by ascorbic acid. Elevated temperatures, the factor accelerating substances' decomposition reaction rate, were used in the tests. The research was conducted at two temperatures at intervals of 10 °C: 80 ± 0.1 and 90 ± 0.1 °C. It was stated that the studied substances' decomposition occurred in accordance with the equation for first-order reactions. The function of the logarithmic concentration (log%C) over time was revealed to be rectilinear. This dependence was used to determine the kinetics of decomposition reaction rate parameters. The stabilization of vitamin C solutions was measured as the time in which 10 % of the substance decomposed at 20 and 0 °C. Addition of Se(IV) or Mg(II) ions significantly increase the stability of ascorbic acid solution (∼34 and ∼16 %, respectively), but Zn(II) causes a significant decrease in stability by ∼23 %. Addition of Mn(II) has no significant influence on vitamin C stability.
Sigma-Aldrich
Sodium selenite, 99%
Sigma-Aldrich
Sodium selenite, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Sodium selenite, γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium selenite, anhydrous, ≥90.0% (RT)
Sigma-Aldrich
Zinc acetate dihydrate, 99.999% trace metals basis
Sigma-Aldrich
Fumaric acid, ≥99.0% (T)
Sigma-Aldrich
Fumaric acid, FCC, FG
Sigma-Aldrich
Manganese(II) sulfate monohydrate, BioReagent, suitable for plant cell culture, suitable for cell culture
Sigma-Aldrich
Manganese sulfate monohydrate, meets USP testing specifications
Sigma-Aldrich
Manganese(II) sulfate monohydrate, puriss., meets analytical specification of Ph. Eur, BP, USP, FCC, 99-100.5% (calc. for dried substance)
Sigma-Aldrich
Zinc acetate dihydrate, reagent grade
Sigma-Aldrich
Manganese(II) sulfate monohydrate, ReagentPlus®, ≥99%
Sigma-Aldrich
Zinc acetate dihydrate, ACS reagent, ≥98%
Supelco
Fumaric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Fumaric acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Zinc acetate dihydrate, puriss. p.a., ACS reagent, ≥99.0% (KT)
Sigma-Aldrich
Zinc acetate dihydrate, puriss., E 650, 99-102%
Sigma-Aldrich
Manganese(II) sulfate monohydrate, ACS reagent, ≥98%