MilliporeSigma
  • Influence of activated charcoal on desorption kinetics and biodegradation of phenanthrene in soil.
Influence of activated charcoal on desorption kinetics and biodegradation of phenanthrene in soil.
Environmental science & technology (2012-10-25)
Angela H Rhodes, Matthew J Riding, Laura E McAllister, Katherine Lee, Kirk T Semple
ABSTRACT
The observed strong sorption of polycyclic aromatic hydrocarbons (PAHs) to black carbon (BC) presents potential implications for PAH bioaccessibility in soils. The effects of BC on the desorption kinetics and mineralization of phenanthrene in four soils was investigated after 1, 25, 50, and 100 d soil-PAH contact time, using sequential hydroxypropyl-β-cyclodextrin (HPCD) extractions in soils amended with 0, 0.1, 1, and 5% (dry wt. soil) activated charcoal (AC, a form of BC). The rapidly (%F(rap)) and slowly (%F(slow)) desorbing phenanthrene fractions and their rate constants were determined using a first-order two-compartment (biphasic) desorption model. A minimum 7.8-fold decrease in %F(rap) occurred when AC was increased from 0 to 5%, with a corresponding increase in %F(slow). Desorption rate constants followed the progression k(rap) (% h(-1)) > k(slow) (% h(-1)) and were in the order of 10(-1) to 10(-2) and 10(-3) to 10(-4), respectively. Linear regressions between %F(rap) and the fractions degraded by a phenanthrene-degrading inoculum (%F(min)) indicated that slopes did not approximate 1 at concentrations greater than 0% AC; %F(min) often exceeded %F(rap), indicating a fraction of sorbed phenanthrene (%F(slow)) remained microbially accessible. Therefore, HPCD-desorption kinetics alone may not be an adequate basis for the prediction of the bioaccessibility of PAHs to microorganisms or bioremediation potential in AC-amended soils.
Sigma-Aldrich
Phenanthrene, sublimed grade, ≥99.5%
Supelco
Phenanthrene, analytical standard, for environmental analysis
Sigma-Aldrich
Phenanthrene, 98%
Supelco
Phenanthrene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland