MilliporeSigma
  • Effects of feeding corn naturally contaminated with aflatoxin B1 and B2 on hepatic functions of broilers.
Effects of feeding corn naturally contaminated with aflatoxin B1 and B2 on hepatic functions of broilers.
Poultry science (2012-10-24)
J Yang, F Bai, K Zhang, S Bai, X Peng, X Ding, Y Li, J Zhang, L Zhao
ABSTRACT
The purpose of this study was to evaluate the effects of feeding corn naturally contaminated with aflatoxin B(1) (AFB(1)) and aflatoxin B(2) (AFB(2)) on serum biochemical parameters, hepatic antioxidant enzyme activities, and pathological lesions of broilers. In total, 1,200 Cobb male broilers were randomly allocated into 5 treatments, with 8 replicates per treatment and 30 birds per replicate, in a 42-d experiment. The dietary treatments were as follows: control, 25, 50, 75, and 100% contaminated corn groups. Results showed that serum aspartate aminotransferase activity in the 75 and 100% contaminated groups were higher than that in the control group on d 21 (P < 0.05). Decreased content of hepatic total protein and increased activities of hepatic glutathione reductase and glutathione-S-transferase were observed as the percentage of contaminated corn increased (P < 0.05). The activity of superoxide dismutase and the content of hepatic malondialdehyde increased when the broilers were fed with more than 50% contaminated corn (P < 0.05). A reduction in glutathione peroxidase level was observed in the AFB(1)- and AFB(2)-contaminated groups on d 21 (P < 0.05). The average pathological lesion scores and apoptosis rate of liver cells increased as the concentration of dietary AFB(1) and AFB(2) increased. Ultrastructural changes were found in the livers of broilers fed 100% contaminated corn. In conclusion, diets containing AFB(1) and AFB(2) could induce pathological lesions in the livers, slightly change the serum biochemical parameters, and damage the hepatic antioxidant functions when the inclusion of AFB(1)- and AFB(2)-contaminated corn reached or exceeded 50%.
Product Description
Aflatoxin B2 solution, 3.80 μg/g in acetonitrile, ERM®, certified reference material
Supelco
Aflatoxin B2 solution, 0.5 μg/mL in acetonitrile, analytical standard
Sigma-Aldrich
Aflatoxin B2