MilliporeSigma
  • Synthesis and characterization of Bi2O3/HSA core-shell nanoparticles for X-ray imaging applications.
Synthesis and characterization of Bi2O3/HSA core-shell nanoparticles for X-ray imaging applications.
Journal of biomedical materials research. Part B, Applied biomaterials (2012-10-24)
Hagit Aviv, Soenke Bartling, Igor Grinberg, Shlomo Margel
ABSTRACT
Bismuth oxide nanoparticles of 12.1 ± 3.0 nm diameter were prepared by thermal decomposition of bismuth acetate dissolved in ethylene glycol in the presence of an oxidizing agent. Functionalization and stabilization of the hydrophobic Bi(2)O(3) nanoparticles was accomplished by coating these core nanoparticles with human serum albumin (HSA), via a precipitation process. The formed Bi(2)O(3)/HSA core-shell nanoparticles were of 15.2 ± 3.5 nm diameter. Elemental analysis measurements indicated that the bismuth weight % of the Bi(2)O(3)/HSA core-shell nanoparticles is 72.9. The crystalline structure of these nanoparticles was examined by XRD. The radiopacity of these nanoparticles was demonstrated in vitro and in vivo by a CT scanner. In ovo and in vivo trials proved the safety of these Bi(2)O(3)/HSA core-shell nanoparticles. In the future, we plan to extend this study particularly for molecular imaging applications.
Sigma-Aldrich
Bismuth(III) oxide, nanopowder, 90-210 nm particle size, 99.8% trace metals basis
Sigma-Aldrich
Bismuth(III) oxide, powder, 99.999% trace metals basis
Sigma-Aldrich
Bismuth(III) oxide, purum, ≥98.0% (KT)
Sigma-Aldrich
Bismuth(III) oxide, ReagentPlus®, powder, 10 μm, 99.9% trace metals basis