MilliporeSigma
  • Prussian blue nanoparticles operate as a new generation of photothermal ablation agents for cancer therapy.
Prussian blue nanoparticles operate as a new generation of photothermal ablation agents for cancer therapy.
Chemical communications (Cambridge, England) (2012-10-24)
Guanglei Fu, Wei Liu, Shanshan Feng, Xiuli Yue
ABSTRACT
Herein, prussian blue nanoparticles, an ancient dye, were explored as a new generation of near-infrared laser-driven photothermal ablation agents for cancer therapy alternative to traditional agents due to their good photothermal efficiency and high photothermal stability but low cost and particularly clinically approved biosafety.
Sigma-Aldrich
Iron(III) ferrocyanide