MilliporeSigma
  • Transition-metal-free synthesis of aryl-substituted tert-butyl ynol ethers through addition/elimination substitution at an sp centre.
Transition-metal-free synthesis of aryl-substituted tert-butyl ynol ethers through addition/elimination substitution at an sp centre.
Chemistry (Weinheim an der Bergstrasse, Germany) (2012-10-23)
Vincent J Gray, Ben Slater, Jonathan D Wilden
ABSTRACT
Nucleophilic attack of an alkoxide on an alkynyl sulfonamide leads to displacement of the sulfonamide at the sp centre and isolation of the ynol ether in good yield in a single operation. The mechanistic pathway has been probed by the use of coordinating additives, (13)C-labelling experiments and ab initio calculations, which indicated that an addition/elimination mechanism is in operation.
Sigma-Aldrich
tert-Butanol, anhydrous, ≥99.5%
Supelco
tert-Butanol, analytical standard
Sigma-Aldrich
tert-Butanol, puriss. p.a., ACS reagent, ≥99.7% (GC)
Sigma-Aldrich
tert-Butanol, ≥99% (GC)
Sigma-Aldrich
tert-Butanol, TEBOL® 99, ≥99.3%
Sigma-Aldrich
tert-Butanol, ACS reagent, ≥99.0%
Sigma-Aldrich
tert-Butanol, suitable for HPLC, ≥99.5%