MilliporeSigma
  • Experimental study of hydrogen bonding potentially stabilizing the 5'-deoxyadenosyl radical from coenzyme B12.
Experimental study of hydrogen bonding potentially stabilizing the 5'-deoxyadenosyl radical from coenzyme B12.
Chemistry (Weinheim an der Bergstrasse, Germany) (2012-10-20)
Peter Friedrich, Ulrich Baisch, Ross W Harrington, Fredrick Lyatuu, Kai Zhou, Felix Zelder, William McFarlane, Wolfgang Buckel, Bernard T Golding
ABSTRACT
Coenzyme B(12) can assist radical enzymes that accomplish the vicinal interchange of a hydrogen atom with a functional group. It has been proposed that the Co-C bond homolysis of coenzyme B(12) to cob(II)alamin and the 5'-deoxyadenosyl radical is aided by hydrogen bonding of the corrin C19-H to the 3'-O of the ribose moiety of the incipient 5'-deoxyadenosyl radical, which is stabilized by 30 kJ mol(-1) (B. Durbeej et al., Chem. Eur. J. 2009, 15, 8578-8585). The diastereoisomers (R)- and (S)-2,3-dihydroxypropylcobalamin were used as models for coenzyme B(12). A downfield shift of the NMR signal for the C19-H proton was observed for the (R)-isomer (δ=4.45 versus 4.01 ppm for the (S)-isomer) and can be ascribed to an intramolecular hydrogen bond between the C19-H and the oxygen of CHOH. Crystal structures of (R)- and (S)-2,3-dihydroxypropylcobalamin showed C19-H⋅⋅⋅O distances of 3.214(7) Å (R-isomer) and 3.281(11) Å (S-isomer), which suggest weak hydrogen-bond interactions (-ΔG<6 kJ mol(-1)) between the CHOH of the dihydroxypropyl ligand and the C19-H. Exchange of the C19-H, which is dependent on the cobalt redox state, was investigated with cob(I)alamin, cob(II)alamin, and cob(III)alamin by using NMR spectroscopy to monitor the uptake of deuterium from deuterated water in the pH range 3-11. No exchange was found for any of the cobalt oxidation states. 3',5'-Dideoxyadenosylcobalamin, but not the 2',5'-isomer, was found to act as a coenzyme for glutamate mutase, with a 15-fold lower k(cat)/K(M) than 5'-deoxyadenosylcobalamin. This indicates that stabilization of the 5'-deoxyadenosyl radical by a hydrogen bond that involves the C19-H and the 3'-OH group of the cofactor is, at most, 7 kJ mol(-1) (-ΔG). Examination of the crystal structure of glutamate mutase revealed additional stabilizing factors: hydrogen bonds between both the 2'-OH and 3'-OH groups and glutamate 330. The actual strength of a hydrogen bond between the C19-H and the 3'-O of the ribose moiety of the 5'-deoxyadenosyl group is concluded not to exceed 6 kJ mol(-1) (-ΔG).
Sigma-Aldrich
Coenzyme B12, ≥97.0%