MilliporeSigma
  • Decontamination and digestion of infectious animal waste using a tissue dissolver in an animal biosafety level 3 facility.
Decontamination and digestion of infectious animal waste using a tissue dissolver in an animal biosafety level 3 facility.
Lab animal (2012-10-20)
Lesley C Homer, Daniel J Fisher, Dennis T Heflin, Kelly Stefano Cole
ABSTRACT
Alkaline hydrolysis-based tissue dissolvers (TDs) are commercially available tools for the digestion and decontamination of infectious animal waste. The authors carried out a series of experiments to verify whether the TD in their facility completely digested animal carcasses and inactivated infectious agents. Using the manufacturer's recommended cycle parameters, the TD inactivated a high concentration of chemically resistant bacterial spores used as a surrogate for the infectious agents in use in the facility. Animal tissues were completely digested into a non-infectious liquid effluent that could be disposed of directly to the sanitary sewer. Reducing the cycle time by 50% still inactivated all spores, although a small amount of tissue remained undigested. The authors recommend that each facility carry out its own experiments to verify the efficacy of a TD before use, given that the design and style of TD as well as the composition of the tissue load is likely to vary between facilities.
Product Description
Supelco
Potassium hydroxide concentrate, 0.1 M KOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis
Sigma-Aldrich
Potassium hydroxide, ≥85% KOH basis, pellets, white
Supelco
Potassium hydroxide solution, volumetric, 8.0 M KOH (8.0N)
Sigma-Aldrich
Potassium hydroxide, tested according to Ph. Eur.
Sigma-Aldrich
Potassium hydroxide, free-flowing, Redi-Dri, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Potassium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, 85-100.5%, pellets
Sigma-Aldrich
Potassium hydroxide, technical, ≥85%, powder
Sigma-Aldrich
Potassium hydroxide, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Potassium hydroxide, pellets, reag. Ph. Eur., ≥85%
Sigma-Aldrich
Potassium hydroxide, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Sigma-Aldrich
Potassium hydroxide, reagent grade, 90%, flakes
Sigma-Aldrich
Potassium hydroxide, puriss. p.a., ≥86% (T), pellets
Sigma-Aldrich
Potassium hydroxide solution, 45 wt. % in H2O