MilliporeSigma
  • Palladium-catalyzed aerobic dehydrogenative aromatization of cyclohexanone imines to arylamines.
Palladium-catalyzed aerobic dehydrogenative aromatization of cyclohexanone imines to arylamines.
Organic letters (2012-10-18)
Alakananda Hajra, Ye Wei, Naohiko Yoshikai
ABSTRACT
Dehydrogenative aromatization of cyclohexanone imines to arylamines has been achieved using a palladium catalyst under aerobic conditions. The reaction is applicable to a variety of imines that are either preformed or generated in situ from cyclohexanone derivatives and aryl or alkylamines.
Product Description
Supelco
Cyclohexanone, Selectophore, for ion-selective electrodes, ≥99.5%
Sigma-Aldrich
Cyclohexanone, 99.8%
Sigma-Aldrich
Cyclohexanone, puriss. p.a., ≥99.5% (GC)
Supelco
Cyclohexanone, analytical standard
Sigma-Aldrich
Cyclohexanone, ReagentPlus®, 99.8%
Sigma-Aldrich
Cyclohexanone, ACS reagent, ≥99.0%