MilliporeSigma
  • Biomarker responses in earthworm Eisenia andrei exposed to pirimiphos-methyl and deltamethrin using different toxicity tests.
Biomarker responses in earthworm Eisenia andrei exposed to pirimiphos-methyl and deltamethrin using different toxicity tests.
Chemosphere (2012-10-16)
Mirna Velki, Branimir K Hackenberger
ABSTRACT
The effects of two widely used insecticides - organophosphate pirimiphos-methyl and pyrethroid deltamethrin - were investigated under laboratory conditions following OECD guidelines using the epigeic earthworm Eisenia andrei as the test organism. The overall aim of this study was to evaluate the effects of these pesticides on molecular biomarkers of earthworm E. andrei using the in vitro, filter paper contact and artificial soil test. In this study for the first time the equivalent concentrations of investigated pesticide applied in different tests were calculated. Although the response of measured molecular biomarkers in different toxicity tests had certain similarities, some distinct differences were also evident. Both pesticides inhibited AChE and CES activities in all three applied toxicity tests; however only in the filter paper test the hormetic effect was recorded. The artificial soil test showed that duration of the exposure significantly changed the effects of the investigated pesticides on CAT and GST activities. Namely, after the initial increase, the prolongation of exposure caused the reduction of the CAT and GST activities. Both pesticides significantly inhibited the efflux pump activity. In the artificial soil test, the significant changes in measured biomarkers after application of doses lower than doses recommended for use in the agriculture indicate that the investigated pesticides could have a harmful effect on earthworms in the context of the realistic environment.
Product Description
Supelco
Pirimiphos-methyl, PESTANAL®, analytical standard