MilliporeSigma
  • Effects of conjugated linoleic acid and high oleic acid safflower oil in the treatment of children with HPV-induced laryngeal papillomatosis: a randomized, double-blinded and crossover preliminary study.
Effects of conjugated linoleic acid and high oleic acid safflower oil in the treatment of children with HPV-induced laryngeal papillomatosis: a randomized, double-blinded and crossover preliminary study.
Lipids in health and disease (2012-10-16)
Louise Louw
ABSTRACT
Surgery is the mainstay therapy for HPV-induced laryngeal papillomatosis (LP) and adjuvant therapies are palliative at best. Research revealed that conjugated-linoleic acid (CLA) may improve the outcome of virally-induced diseases. The effects of Clarinol™ G-80 (CLA) and high oleic safflower oil (HOSF) on children with LP (concomitant with surgery) were evaluated. A randomized, double-blinded, crossover and reference-oil controlled trial was conducted at a South African medical university. Study components included clinical, HPV type/load and lymphocyte/cytokine analyses, according to routine laboratory methods. Overall: ten children enrolled; eight completed the trial; five remained randomized; seven received CLA first; all treatments remained double-blinded. Children (4 to 12 years) received 2.5 ml p/d CLA (8 weeks) and 2.5 ml p/d HOSF (8 weeks) with a washout period (6 weeks) in-between. The one-year trial included a post-treatment period (30 weeks) and afterwards was a one-year follow-up period. Changes in numbers of surgical procedures for improved disease outcome, total/anatomical scores (staging system) for papillomatosis prevention/viral inhibition, and lymphocyte/cytokine counts for immune responses between baselines and each treatment/end of trial were measured. After each treatment all the children were in remission (no surgical procedures); after the trial two had recurrence (surgical procedures in post-treatment period); after the follow-up period three had recurrence (several surgical procedures) and five recovered (four had no surgical procedures). Effects of CLA (and HOSF to a lesser extent) were restricted to mildly/moderately aggressive papillomatosis. Children with low total scores (seven/less) and reduced infections (three/less laryngeal sub-sites) recovered after the trial. No harmful effects were observed. The number of surgical procedures during the trial (n6/available records) was significantly lower [(p 0.03) (95% CI 1.1; 0)]. Changes in scores between baselines and CLA treatments (n8) were significantly lower: total scores [(p 0.02) (95% CI -30.00; 0.00)]; anatomical scores [(p 0.008) (95% CI -33.00: -2.00)]. Immune enhancement could not be demonstrated. These preliminary case and group findings pave the way for further research on the therapeutic potential of adjuvant CLA in the treatment of HPV-induced LP.
Sigma-Aldrich
Safflower seed oil from Carthamus tinctorius seed