MilliporeSigma
Hexathienocoronenes: synthesis and self-organization.
Journal of the American Chemical Society (2012-10-16)
Long Chen, Sreenivasa R Puniredd, Yuan-Zhi Tan, Martin Baumgarten, Ute Zschieschang, Volker Enkelmann, Wojciech Pisula, Xinliang Feng, Hagen Klauk, Klaus Müllen
ABSTRACT
Here we report hexathienocoronenes (HTCs), fully thiophene-annelated coronenes in which six double bonds in the periphery are thieno-fused. The derivatives tetrasubstituted with hexyl and dodecyl chains show a phase formation that strongly depends on the chain length. HTCs are remarkably stronger donors than the known thiophene-annelated coronenes but do not readily assemble into well-ordered films when deposited from the vapor phase. Thus, thin-film transistors fabricated by vacuum deposition have only modest field-effect mobilities of 0.002 cm(2) V(-1) s(-1).
Sigma-Aldrich
Coronene, purified by sublimation, 99%
Coronene, BCR®, certified reference material
Sigma-Aldrich
Coronene, 97%