MilliporeSigma
Sargassum myriocystum mediated biosynthesis of gold nanoparticles.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2012-10-13)
T Stalin Dhas, V Ganesh Kumar, L Stanley Abraham, V Karthick, K Govindaraju
ABSTRACT
Functionalized metal nanoparticles are unique in nature and are being developed for its specificity in drug targeting. In the present study, aqueous extract of Sargassum myriocystum is used for the biosynthesis of gold nanoparticles (AuNPs) by the reduction of chloroauric acid. The formation of nanoparticles reaction was complete within 15 min at 76 °C. The size, shape and elemental analysis of AuNPs were carried out using UV-visible absorption spectroscopy, FT-IR, TEM, SEM-EDAX, and XRD analysis. The newly formed AuNPs are stable, well-defined, polydispersed (triangular and spherical) and crystalline with an average size of 15 nm. The biomolecule involved in stabilizing AuNPs was identified using GC-MS.
Sigma-Aldrich
Sodium tetrachloroaurate(III) dihydrate, 99%
Sigma-Aldrich
Gold(III) chloride solution, 99.99% trace metals basis, 30 wt. % in dilute HCl
Sigma-Aldrich
Potassium gold(III) chloride, 99.995% trace metals basis
Sigma-Aldrich
Potassium gold(III) chloride, 98%