MilliporeSigma
  • Empathy in electrodermal responsive and nonresponsive patients with schizophrenia.
Empathy in electrodermal responsive and nonresponsive patients with schizophrenia.
Schizophrenia research (2012-10-13)
Satoru Ikezawa, Silvia Corbera, Jiacheng Liu, Bruce E Wexler
ABSTRACT
Skin conductance response (SCR) to emotion-evoking stimuli has in previous studies suggested the existence of two subgroups of schizophrenia patients. One is characterized by absent SCR and the other by heightened or non-habituating SCR. These subgroups have also been shown to differ in symptoms, prognosis and social integration. The present project compared social cognition in the two subgroups. SCR from 28 patients with schizophrenia and 24 matched healthy controls was measured while they watched emotion-evoking and neutral video tapes. Assessments of symptoms, neurocognition, social cognition, and social function were also performed. Event related potentials (ERP) were recorded in response to pictures of people experiencing pain or not. Subjects were divided into "SCR non-responder" and "SCR responder" groups based on SCR frequency. Schizophrenia SCR responders had significantly higher self-reported personal distress in response to others in distress and lower P300 ERP responses to others in pain than schizophrenia SCR non-responders and healthy controls. SCR responsiveness is a potential marker of subgroups of patients with schizophrenia that differ in pathophysiology, function and prognosis.
Product Description
Chlorpromazine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Chlorpromazine hydrochloride, VETRANAL®, analytical standard
Sigma-Aldrich
Chlorpromazine hydrochloride, meets USP testing specifications
Sigma-Aldrich
Chlorpromazine hydrochloride, ≥98% (TLC)
Supelco
Chlorpromazine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®