MilliporeSigma
  • Three-dimensional morphology of microdamage in peri-screw bone: a scanning electron microscopy of methylmethacrylate cast replica.
Three-dimensional morphology of microdamage in peri-screw bone: a scanning electron microscopy of methylmethacrylate cast replica.
Microscopy and microanalysis : the official journal of Microscopy Society of America, Microbeam Analysis Society, Microscopical Society of Canada (2012-10-11)
Lei Wang, Jin Shao, Tingjun Ye, Lianfu Deng, Shijing Qiu
ABSTRACT
Screw implantation inevitably causes microdamage in surrounding bone. However, little is known about the detailed characteristics of microdamage in peri-screw bone. In this study, we developed a method to construct microdamage cast with methylmethacrylate (MMA) and observed the cast using scanning electron microscopy (SEM). In basic fuchsin stained bone sections observed by bright-field and fluorescence microscopy, diffuse damage, cross-hatched damage, and linear cracks were all presented in peri-screw bone. Using MMA casting/SEM method, we found numerous densely packed microcracks in the areas with diffuse damage. The osteocyte canaliculi and the microcracks consisting of diffuse damage had a similar diameter (or width), usually <0.5 μm, but their morphology was largely different. In the area with cross-hatched damage, the orientation of microcracks was similar to that in diffuse damage, but the number was significantly decreased. Many microcracks were thicker than 1 μm and associated with a rough surface. Large linear cracks (∼10 μm in diameter) occurred in different areas. Plenty of microcracks were present on the surface of some linear cracks. In conclusion, the MMA casting/SEM method can demonstrate the three-dimensional morphology of different types of microdamage, particularly the microcracks in diffuse damage, which are unable to be shown by light microscopy.
Sigma-Aldrich
Methyl methacrylate, 99%, stabilized
Supelco
Methyl methacrylate solution, certified reference material, 1000 μg/mL in methanol
Sigma-Aldrich
Methyl methacrylate, contains ≤30 ppm MEHQ as inhibitor, 99%