MilliporeSigma
  • Interactions with β-cyclodextrin as a way for encapsulation and separation of camphene and fenchene.
Interactions with β-cyclodextrin as a way for encapsulation and separation of camphene and fenchene.
Carbohydrate polymers (2012-10-10)
Magdalena Ceborska, Monika Asztemborska, Roman Luboradzki, Janusz Lipkowski
ABSTRACT
The separation of isomeric monoterpenes, camphene and fenchene by complexation with β-cyclodextrin is presented. Both of the monoterpenes form complexes with β-cyclodextrin (as shown by both gas chromatography and (1)H NMR) with similar stability constants nevertheless it is possible to separate them by re-crystallization. The crystal structure of β-cyclodextrin with fenchene was also studied by X-ray diffraction.
Product Description
Supelco
Camphene, analytical standard
Sigma-Aldrich
Camphene, 95%