MilliporeSigma
Helquat-induced chiroselective aggregation of Au NPs.
Nano letters (2012-10-10)
Dora Balogh, Zhanxia Zhang, Alessandro Cecconello, Jan Vavra, Lukas Severa, Filip Teply, Itamar Willner
ABSTRACT
Au nanoparticles (NPs) are functionalized with chiral (R) or (S) binaphthol phenylboronic acid ligands, (1a) or (1b). The (R)- or (S)-binaphthol phenylboronic acid ligands form donor-acceptor complexes with the chiral dicationic helicene, helquat (P)-HQ(2+) or (M)-HQ(2+), (2a) or (2b). The association constants between (1a)/(2a) and (1a)/(2b) correspond to (7.0 ± 0.5) × 10(5) M(-1) and (2.5 ± 0.3) × 10(5) M(-1), respectively, whereas the association constants between (1b)/(2b) and (1b)/(2a) correspond to (4.0 ± 0.5) × 10(5) M(-1) and (1.8 ± 0.3) × 10(5) M(-1), respectively. Chiroselective aggregation of chiral binaphthol phenylboronic acid-capped Au NPs triggered by the chiral helquats, is demonstrated.
Sigma-Aldrich
Phenylboronic acid, 95%
Sigma-Aldrich
Phenylboronic acid, purum, ≥97.0% (HPLC)