MilliporeSigma
Cocaine-levamisole thrombotic vasculopathy.
Seminars in thrombosis and hemostasis (2012-10-09)
Irene A Dy, Peter H Wiernik
ABSTRACT
The syndrome of cocaine-levamisole thrombotic vasculopathy is a tetrad of cutaneous manifestations, arthralgia, leukopenia, and positive antineutrophil cytoplasmic antigen (ANCA). Cocaine is cut with levamisole to potentiate its euphoric effects. However, along with this are the adverse reactions of levamisole such as fatal agranulocytosis, flu-like symptoms, and cutaneous thrombotic vasculopathy. High index of suspicion is needed for its diagnosis. This article will review on its toxicities, especially its prothrombotic effects.
Product Description
Supelco
(−)-Levamisole hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Levamisol hydrochloride, VETRANAL®, analytical standard
Levamisole hydrochloride for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(−)-Tetramisole hydrochloride, ≥99% (GC)
Levamisole hydrochloride, European Pharmacopoeia (EP) Reference Standard