MilliporeSigma
  • Synthesis of hollow polymeric nanoparticles for protein delivery via inverse miniemulsion periphery RAFT polymerization.
Synthesis of hollow polymeric nanoparticles for protein delivery via inverse miniemulsion periphery RAFT polymerization.
Chemical communications (Cambridge, England) (2012-10-09)
Robert H Utama, Yi Guo, Per B Zetterlund, Martina H Stenzel
ABSTRACT
Hollow polymeric nanoparticles with a hydrophilic liquid core have been synthesized in a one-pot approach via a novel inverse miniemulsion periphery RAFT polymerization process. Successful encapsulation and release of a model protein is reported as a potential application.
Sigma-Aldrich
Ethylene glycol dimethacrylate, 98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor