MilliporeSigma
  • Evaluation of the partial renewal of in situ phytoplankton microcosms and application to the impact assessment of bentazon and dimethenamid.
Evaluation of the partial renewal of in situ phytoplankton microcosms and application to the impact assessment of bentazon and dimethenamid.
Marine pollution bulletin (2012-10-09)
Denis de la Broise, Sabine Stachowski-Haberkorn
ABSTRACT
Microcosms, each consisting of 2L natural surface seawater maintained in 2.3-L glass bottles, were immersed at a depth of 6m. The renewal of 10% of microcosm volumes was carried out every other day. Phytoplankton-containing seawater was used for renewal (previously filtered through 25-, 50- or 200-μm cut-off). Phytoplankton community pigment analysis (by HPLC) and flow cytometry analysis were performed. After 13 days, data exhibited phytoplankton characteristics in microcosms in the same range as that of the natural surrounding sea water over the same period. Furthermore, in these microcosms, a negative correlation was observed between the filtration cut-off used for renewal water, and the total cell count. Herbicides were tested as commercial mixtures at 1, 10 and 100 μgL(-1) active substance. Both Frontier® (dimethenamid) and Basamais® (bentazon) induced significant modifications of the phytoplankton populations at every concentration tested. Such results suggest a possible disturbance in polluted coastal areas.
Product Description
Supelco
Bentazon, PESTANAL®, analytical standard