MilliporeSigma
  • Comparative clinical study of the effectiveness of different dental bleaching methods - two year follow-up.
Comparative clinical study of the effectiveness of different dental bleaching methods - two year follow-up.
Journal of applied oral science : revista FOB (2012-10-04)
Rafael Francisco Lia Mondelli, Juliana Felipi David E Góes de Azevedo, Ana Carolina Francisconi, Cristiane Machado de Almeida, Sérgio Kiyoshi Ishikiriama
ABSTRACT
This study evaluated color change, stability, and tooth sensitivity in patients submitted to different bleaching techniques. In this study, 48 patients were divided into five groups. A half-mouth design was conducted to compare two in-office bleaching techniques (with and without light activation): G1: 35% hydrogen peroxide (HP) (Lase Peroxide - DMC Equipments, São Carlos, SP, Brazil) + hybrid light (HL) (LED/Diode Laser, Whitening Lase II DMC Equipments, São Carlos, SP, Brazil); G2: 35% HP; G3: 38% HP (X-traBoost - Ultradent, South Jordan UT, USA) + HL; G4: 38% HP; and G5: 15% carbamide peroxide (CP) (Opalescence PF - Ultradent, South Jordan UT, USA). For G1 and G3, HP was applied on the enamel surface for 3 consecutive applications activated by HL. Each application included 3x3' HL activations with 1' between each interval; for G2 and G4, HP was applied 3x15' with 15' between intervals; and for G5, 15% CP was applied for 120'/10 days at home. A spectrophotometer was used to measure color change before the treatment and after 24 h, 1 week, 1, 6, 12, 18 and 24 months. A VAS questionnaire was used to evaluate tooth sensitivity before the treatment, immediately following treatment, 24 h after and finally 1 week after. Statistical analysis did not reveal any significant differences between in-office bleaching with or without HL activation related to effectiveness; nevertheless the time required was less with HL. Statistical differences were observed between the results after 24 h, 1 week and 1, 6, 12, 18 and 24 months (intergroup). Immediately, in-office bleaching increased tooth sensitivity. The groups activated with HL required less application time with gel. All techniques and bleaching agents used were effective and demonstrated similar behaviors.
Sigma-Aldrich
Urea hydrogen peroxide, 97%
Sigma-Aldrich
Hydrogen peroxide–Urea adduct, powder, 15-17% active oxygen basis