MilliporeSigma
  • Intraocular pressure-lowering effects of commonly used fixed-combination drugs with timolol: a systematic review and meta-analysis.
Intraocular pressure-lowering effects of commonly used fixed-combination drugs with timolol: a systematic review and meta-analysis.
PloS one (2012-10-03)
Jin-Wei Cheng, Shi-Wei Cheng, Lian-Di Gao, Guo-Cai Lu, Rui-Li Wei
ABSTRACT
The first goal of medical therapy in glaucoma is to reduce intraocular pressure (IOP), and the fixed-combination medications are needed to achieve sufficiently low target IOP. The aim of this systematic review and meta-analysis is to evaluate IOP-lowering effect of the commonly used fixed-combination drugs containing 0.5% timolol. Pertinent publications were identified through systematic searches. Over 85% of the patients had to be diagnosed with primary open-angle glaucoma (POAG) or ocular hypertension (OHT). Forty-one randomized clinical trials were included in the meta-analysis. The main efficacy measures were the absolute and relative values of mean diurnal IOP reduction, and the highest and lowest IOP reductions on the diurnal IOP curve. The pooled 1- to 3-month IOP-lowering effects after a medicine-free washout period was calculated by performing meta-analysis using the random effects model, and relative treatment effects among different fixed combinations were assessed using a mixed-effects meta-regression model. The relative reductions for mean diurnal IOP were 34.9% for travoprost/timolol, 34.3% for bimatoprost/timolol, 33.9% for latanoprost/timolol, 32.7% for brinzolamide/timolol, 29.9% for dorzolamide/timolol, and 28.1% for brimonidine/timolol. For the highest IOP decrease, relative reductions ranged from 31.3% for dorzolamide/timolol to 35.5% for travoprost/timolol; for the lowest IOP decrease, those varied from 25.9% for dorzolamide/timolol to 33.1% for bimatoprost/timolol. Both latanoprost/timolol and travoprost/timolol were more effective in lowering mean diurnal IOP than brimonidine/timolol (WMD: 5.9 and 7.0) and dorzolamide/timolol (WMD: 3.8 and 3.3). All six commonly used fixed-combination drugs containing timolol can effectively lower IOP in patients with POAG and OHT, and both latanoprost/timolol and travoprost/timolol might achieve better IOP-lowering effects among the six fixed-combination agents.
Sigma-Aldrich
Timolol maleate salt, ≥98% (TLC), powder