MilliporeSigma
Histochemical detection of lipid droplets in cultured cells.
Methods in molecular biology (Clifton, N.J.) (2012-10-03)
Michitaka Suzuki, Yuki Shinohara, Toyoshi Fujimoto
ABSTRACT
Cells store excess lipid as esters in the form of triglycerides and cholesterol esters. Most lipid esters are compartmentalized in globular structures called lipid droplets. Here we describe several methods of detecting lipid droplets by fluorescence microscopy. Lipid droplets can be visualized either by staining the lipid ester core using fluorescent dyes or by labeling lipid droplet-specific proteins using antibodies. The intracellular distribution of lipid droplets can be analyzed without much difficulty by these methods, but care must be taken to avoid certain pitfalls.
Product Description
Supelco
Oil Red O, BioReagent, suitable for electrophoresis
Sigma-Aldrich
Oil Red O, certified by the Biological Stain Commission
Sigma-Aldrich
Oil Red O solution, 0.5% in isopropanol
Sigma-Aldrich
Oil Red O solution, 0.5% in propylene glycol
Sigma-Aldrich
Nile Red, Technical grade
Sigma-Aldrich
Nile Red, for microscopy
Sigma-Aldrich
Nile Red, BioReagent, suitable for fluorescence, ≥97.0% (HPLC)